Learn More
CD3e Armenian Hamster, PE, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB2-PE-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15991638
  • 63.21 € / 25µg
Estimated Shipment: 01-12-2021
Specifications
CD3e
145-2C11
PE
XP_006510029
Armenian Hamster
Protein A/G Purified
RUO
Primary
12501
Flow Cytometry
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd3e
IgG
25 μg
Store at 2-4°C.