missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Armenian Hamster, PerCP-Cy5.5, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
94.25 € - 252.00 €
Specifications
Antigen | CD3e |
---|---|
Clone | 145-2C11 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971638
|
EnquireBio
QAB2-PCP55-25UG |
25 μg |
94.25 €
25µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961638
|
EnquireBio
QAB2-PCP55-100UG |
100 μg |
252.00 €
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3e | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12501 | |
IgG | |
Protein A/G Purified |
145-2C11 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
XP_006510029 | |
Cd3e | |
Primary | |
Store at 2-4°C. |