missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Armenian Hamster, Qfluor 630, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
203.00 € - 385.00 €
Specifications
Antigen | CD3e |
---|---|
Clone | 145-2C11 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951648
|
EnquireBio
QAB2-QF630-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971648
|
EnquireBio
QAB2-QF630-500UG |
500 μg |
276.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961648
|
EnquireBio
QAB2-QF630-1MG |
1 mg |
385.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3e | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12501 | |
IgG | |
Protein A/G Purified |
145-2C11 | |
Flow Cytometry | |
Qfluor 630 | |
RUO | |
XP_006510029 | |
Cd3e | |
Primary | |
Store at 2-4°C. |