missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse, APC, Clone: RPA-T4, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB12-APC-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961508

  • 78.70 € / 25 tests
Estimated Shipment: 20-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD4
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
Protein A/G Purified
RUO
920
IgG1 κ
Flow Cytometry
RPA-T4
APC
NP_001181945
Mouse
25 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers