missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat, PE, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
82.15 € - 94.25 €
Specifications
Antigen | CD45 |
---|---|
Clone | 30-F11 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971968
|
EnquireBio
QAB40-PE-25UG |
25 μg |
82.15 €
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961968
|
EnquireBio
QAB40-PE-100UG |
100 μg |
94.25 €
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD45 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
19264 | |
IgG2b κ | |
Protein A/G Purified |
30-F11 | |
Flow Cytometry | |
PE | |
RUO | |
NP_001255215 | |
Ptprc | |
Primary | |
Store at 2-4°C. |