missing translation for 'onlineSavingsMsg'
Learn More

CD45RO Mouse, PE, Clone: UCHL1, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB44-PE-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15992048

  • 95.36 € / 25 tests

Estimated Shipment: 01-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD45RO
UCHL1
PE
NP_002829
Mouse
Protein A/G Purified
RUO
Primary
5788
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
IgG2a kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates