Learn More
CD68/SR-D1 Antibody (FA-11), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-33337JF646
544.85 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18799872
  • 682.00 € / 0.10mL
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Flow (Intracellular), Single Cell Western
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD68/SR-D1
Monoclonal
Janelia Fluor 646
CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1
This CD68/SR-D1 Antibody (FA-11) was developed against purified ConA acceptor glycoproteins from the P815 cell line.
0.1 mL
Cell Biology, Cytokine Research, Immunology, Microglia Markers
968
Store at 4°C in the dark.
IgG2a
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
FA-11
50mM Sodium Borate
Rat
Protein G purified
RUO
Primary
Mouse
Purified