missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 / MIC2 Rabbit, Clone: MIC2/1495R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | CD99 / MIC2 |
---|---|
Clone | MIC2/1495R |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930478
|
EnquireBio
4267-RBM5-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920478
|
EnquireBio
4267-RBM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD99 / MIC2 | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
4267 | |
Recombinant human full-length MIC2 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
27-32kDa |
MIC2/1495R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001308299 | |
CD99 | |
IgG κ | |
Protein A/G Purified | |
Cell surface | |
MOLT-4 cells. Pancreas or Ewing s sarcoma |