missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 / MIC2 Rabbit, Clone: MIC2/1495R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 4267-RBM5-P1
Additional Details : Weight : 0.01000kg
Specifications
CD99 / MIC2 | |
Cell surface | |
MIC2/1495R | |
Unconjugated | |
NP_001308299 | |
Rabbit | |
27-32kDa | |
Protein A/G Purified | |
RUO | |
4267 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
CD99 | |
Recombinant human full-length MIC2 protein | |
MOLT-4 cells. Pancreas or Ewing s sarcoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |