missing translation for 'onlineSavingsMsg'
Learn More

CDCA2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA549909

Additional Details : Weight : 0.01000kg

Product Code. 15603845

  • 472.00 € / 100µL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody detects endogenous levels of total CDCA2 protein.

CDCA2 Polyclonal antibody specifically detects CDCA2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CDCA2
Polyclonal
Unconjugated
CDCA2
2610311M19Rik; AI586158; CDCA2; cell division cycle associated 2; cell division cycle-associated protein 2; FLJ25804; MGC129906; MGC129907; PPP1R81; protein phosphatase 1, regulatory subunit 81; recruits PP1 onto mitotic chromatin at anaphase protein; Repo-Man
Rabbit
Antigen affinity chromatography
RUO
157313
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q69YH5
CDCA2
Synthesized peptide derived from C-terminal of human CDCA2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers