missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 95%, Thermo Scientific™

158.00 € - 592.00 €

Chemical Identifiers

CAS 78439-06-2
Molecular Formula C22H32N6O12S2
Molecular Weight (g/mol) 636.648
MDL Number MFCD00153936
InChI Key NMVPEQXCMGEDNH-TZVUEUGBSA-N
PubChem CID 131673973
ChEBI CHEBI:3509
IUPAC Name 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
SMILES CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457829
SDS View Documents
Thermo Scientific Alfa Aesar
J66460.03
1g
158.00 €
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15467829
SDS View Documents
Thermo Scientific Alfa Aesar
J66460.06
5g
592.00 €
5g
View alternative products
Save Up to 
»
Estimated Shipment: 18-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Sparingly soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Chemical Identifiers

78439-06-2
636.648
NMVPEQXCMGEDNH-TZVUEUGBSA-N
CHEBI:3509
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
C22H32N6O12S2
MFCD00153936
131673973
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate

Specifications

78439-06-2
C22H32N6O12S2
Sparingly soluble in aqueous solution
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
636.648
CHEBI:3509
95%
White
MFCD00153936
NMVPEQXCMGEDNH-TZVUEUGBSA-N
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
131673973
636.65
Ceftazidime pentahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P363-P501c

H317-H319-H334

missing translation for 'rtecsNumber' : UU2230000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.