Learn More
Thermo Scientific™ Cel-Line Epoxy Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Made of extra-white soda-lime glass with very low iron content. Clean, flat and suitable for immediate use
Brand:  Thermo Scientific™ X8X30#341ABCE
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15393352
  • 67.05 € / Pack of 72
Call for latest local availability
01 885 5854
Add to basket
Description
Description
  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
Specifications
Specifications
75mm
1mm
20mm
1
25mm
Black
Diagnostic Slide
Videos