missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celite™ S, filter aid, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Fiberboard Box
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Gray |
Quantity | 5 kg |
Packaging | Fiberboard Box |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Celite™ S |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'