missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Centerfeed roll, Tork advanced wiper 420, blue
Brand: Manutan A333337
Code : Z3
Additional Details : Weight : 0.01000kg
Description
Two ply multi-purpose paper ideal for mopping up liquids and gentle to frequently washed hands. Fits Standard Tork Centrefeed Wiper Roll Dispenser. Food contact approved.- Blue
- Roll length : 157m
- Width of roll/sheet : 200mm
- Number of sheets : 450