missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Centrifuge Adapter for Hettich Angle Rotors

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 1463

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10042683

  • 35.65 € / Each
Estimated Shipment: 09-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615
Specifications

Specifications

Centrifuge Adapter
50 mL
1
Round Bottom
For Hettich Rotors no. 1620A, 1720, 1792, 4246, 4794, 5615
1
Videos
SDS
Documents
Certificates
Special Offers

Special Offers