missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Catalog Number 15415085
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
1306-38-3
CeO2
172.114
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O
This item is not returnable. View return policy
1306-38-3
CeO2
172.114
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O

The addition of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures. It is used as an additive in glass industry. Cerium dioxide is mainly used as such or in formulation as a high precision polishing agent for glass products, as catalyst or catalyst support. Polishing and decolorizing glass; opacifier in vitreous enamels and photochromic glasses; heat-resistant alloy coatings; as a cracking catalyst; as a catalyst for automobile emission control; in ceramic coatings; in phosphors; in cathodes; in capacitors; in semiconductors; in refractory oxides; gemstone polishing, as raw material for the production of glass and ceramics as well as additives in paints and varnishes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red to Brown
pH 8.0
Physical Form Liquid
Quantity 100 g
Solubility Information Very low water solubility (<0.123 ?g/L at 20°C) and soluble in concentrated mineral acid.
Formula Weight 172.12
Odor Odorless
Density 1.23 g/mL
Chemical Name or Material Cerium(IV) oxide, 20% in H2O, nanoparticle dispersion

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'