missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Melting Point | >400°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 99.4% min. (By difference) |
Linear Formula | Cs2CO3 |
Fieser | 07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73 |
Merck Index | 15,2009 |
Solubility Information | Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether |
IUPAC Name | dicesium;carbonate |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Cesium carbonate
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P263-Avoid contact during pregnancy/while nursing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'