missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 11944211
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
7789-17-5
CsI
259.81
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
[I-].[Cs+]
7789-17-5
CsI
259.81
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
[I-].[Cs+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Boiling Point 1280.0°C
Melting Point 626.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99.999%
Linear Formula CsI
Merck Index 15,2013
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone
IUPAC Name caesium(1+) iodide
Formula Weight 259.8
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Cesium iodide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cesium iodide
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'