Learn More
Cetuximab Mouse, Biotin, UltraLys, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
Brand:  ACROBiosystems CEB-BY31-200ug
 View more versions of this product
Product Code. 16437409
  • 1805.00 € / 200µg
Description
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab, recommended for PK/PD.
Specifications
Specifications
Cetuximab
Biotin
200 μg
Primary
Monoclonal
Hybridoma
RUO
-20°C