missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetuximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
337.00 € - 2465.00 €
Specifications
| Antigen | Cetuximab |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Hybridoma |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16417419
|
ACROBiosystems
CEB-Y31-100ug |
100 μg |
337.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16427419
|
ACROBiosystems
CEB-Y31-1mg |
1 mg |
2465.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab (Non-Neutralizing)Specifications
| Cetuximab | |
| Unconjugated | |
| RUO | |
| -20°C |
| Monoclonal | |
| Hybridoma | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title