missing translation for 'onlineSavingsMsg'
Learn More

CFTR Mouse, Clone: CFTR/1785, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1080-MSM5-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989757

  • 459.10 € / 100µg

Estimated Shipment: 01-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CFTR
Cell Surface and Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
CFTR
Recombinant human CFTR fragment (aa258-385)
165-170kDa
Protein A/G Purified
RUO
Primary
1080
Immunohistochemistry
CFTR/1785
Unconjugated
NP_011514056
Mouse
IgG2b
MOLT-4 cells. Pancreas, Kidney or Placenta.
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates