missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Technical, ActIVated, Acid Washed, Fisher Chemical
Click to view available options
250g, Plastic powder jar
Description
250g, Plastic powder jar
Specifications
CAS | 7440-44-0 |
Melting Point | 3652°C |
Color | Black |
Packaging | Plastic powder jar |
Molecular Formula | C |
MDL Number | 133992 |
Quantity | 250 g |
Formula Weight | 12g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'