missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Technical, Powder, Wood, Fisher Chemical
Click to view available options
1kg, Plastic powder jar
Description
1kg, Plastic powder jar
Specifications
CAS | 7440-44-0 |
Color | Black |
Packaging | Plastic powder jar |
Molecular Formula | C |
MDL Number | 133992 |
Quantity | 1 kg |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'