Learn More
Hach Lange™ Chemical Oxygen Demand Cuvette Test
Pre-dosed LCK cuvette test reagents are easy to use and promote safety. Hach Lange™ Chemical Oxygen Demand Cuvette Test are environmentally friendly because they use less chemicals.
Brand: Hach Lange™ LCK014
Additional Details : CAS Number : 7664-93-9 Weight : 0.32400kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
- Precise and reliable measurement values
- Maximum safety for users, due to the closed system and low reagent amounts
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of color-coding
Order Info
dimRange
Specifications
ISO 6060-1989, DIN 38409-H41-H44 | |
1000 to 10000mg/L O2 | |
Dichromate |
25 | |
15°C to 25°C | |
Chemical Oxygen Demand Cuvette Test |
Safety and Handling
- CSB/COD/DCO
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Respiratory sensitiser Category 1A
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H290-May be corrosive to metals.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid, potassium dichromate, Mercury(II) sulfate, Silver sulfate