missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JULABO™ Refrigerators for Chemicals, KRC Series
Julabo™ Refrigerators for Chemicals is used for storing and cooling of chemicals and hazardous substances. The spark free interior prevents damage caused by spilled or evaporating chemicals.
Brand: JULABO™ 8800705
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- With spark free interior
- Storing and cooling of hazardous substances
- Self-protecting control circuit
- Digital temperature display (LED)
- Overload protection for cooling compressor with test button
- Cut-off in case of disturbance with optical alarm signal
- Working and safety sensors are protected against short circuits and disconnection
Specifications
Refrigerator for chemicals | |
-2°C to +12°C | |
PTC | |
2 | |
Designed for storing and cooling hazardous substances | |
KRC50 | |
64 x 55 x 63 cm | |
41.5 cm | |
230 V 50 Hz | |
23 kg |
68 L | |
0.1°C | |
±1°C | |
LED | |
1 Support rack | |
29 x 42 x 44 cm | |
39 cm | |
230 V | |
50 Hz | |
32 kg |