missing translation for 'onlineSavingsMsg'
Learn More

3M™ Chemical Absorbent Rolls

Relatively inert and powerful enough to handle dilute acids, bases, solvents, oils and many other hazardous liquid spills. 3M™ Chemical Absorbent Rolls are flexible enough to be adapted to a wide range of sizes.

Brand:  3M™ 7000088218

Code : 04

Additional Details : Weight : 4.52250kg

Product Code. 12352459

  • 650.00 € / Pack of 2
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Provides large area coverage
  • Suitable for a wide range of liquids
  • High absorption capacity minimizes amount of waste
Specifications

Specifications

Chemical Sorbent Roll
2/Cs.
330 mm x 30 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers