Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Erlenmeyer Flask
Chemware™ PFA Graduated Erlenmeyer Flask is chemically inert for ultra-pure applications.
119.00 € - 274.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11724628
|
Saint Gobain Life Sciences
D1069075 |
100 mL |
119.00 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
11724738
|
Saint Gobain Life Sciences
D1069076 |
200 mL |
204.00 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
11734738
|
Saint Gobain Life Sciences
D1069077 |
300 mL |
274.00 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
Description
- Graduated
- Transparent for easy viewing
- Non-stick PFA surface-easy to clean
- Can be used in a microwave
Specifications
PFA |