missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Chitosan, molecular weight: 100,000-300,000
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Chitosan is a naturally derived polymer formed from the deacetylation of chitin
- It is an alkaline polysaccharide with moisturizing and adsorption properties
- Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups
Application
- Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
Specifications
Color | Beige-Yellow to White |
Physical Form | Powder or Granules |
Ash | 2% max. |
Quantity | 50 g |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Water | 10% max. (K.F.) |
Chemical Name or Material | Chitosan, Molecular Weight: 100, 000 to 300, 000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'