missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloral hydrate, 98.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Melting Point | 57.0°C |
Boiling Point | 97.0°C |
Assay Percent Range | 98.5% |
Packaging | Glass bottle |
Linear Formula | Cl3CCH(OH)2 |
Quantity | 50 g |
Merck Index | 15, 2071 |
Solubility Information | Solubility aq. soln.: 660 g/100 mL. Other solubilities: soluble in chloroform, alcohol and ether, freely soluble in acetone and methyl ethyl ketone, sparingly soluble in petroleum ether, benzene,, carbon tetrachloride, toluene and turpentine |
Formula Weight | 165.4 |
Chemical Name or Material | Chloral hydrate |
Sécurité et traitement
Identifiant produit
- Chloral hydrate
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'