missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7077866
Click to view available options
Quantity:
1 g
5 g
55-56-1
C22H30Cl2N10
505.45
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
9552079
CHEBI:3614
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl
55-56-1
C22H30Cl2N10
505.45
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
9552079
CHEBI:3614
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 134.0°C to 136.0°C
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 505.45
Percent Purity 98%
Chemical Name or Material Chlorhexidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'