Learn More
Sherwood Scientific™ Chloride Standard Solution
Brand:  Sherwood Scientific™ 00156202
Code : 04
Additional Details : Weight : 0.74340kg
Product Code. 12826823
  • 141.00 € / Pack of 6
Estimated Shipment: 29-04-2024
to see stock.
Specifications
NIST NaCl SRM 919b
100 mmol/L
6/Pk.
100 mL