missing translation for 'onlineSavingsMsg'
Learn More

chlorodimethylphenylsilane, 95%

Product Code. 10647753 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10647753

Brand: Thermo Scientific Acros 151071000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-33-2 | C8H11ClSi | 170.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chloro-dimethyl-phenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0320g/mL
Boiling Point 80°C to 84°C (16.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5Si(CH3)2Cl
Refractive Index 1.5060 to 1.51
Quantity 100 g
Fieser 02,278
Specific Gravity 1.032
Solubility Information Solubility in water: may decompose
Formula Weight 170.72
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chlorodimethylphenylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.