missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, for HPLC, stabilized with amylene, Thermo Scientific Chemicals

Catalog Number p-217331
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
67-66-3, 513-35-9
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
ClC(Cl)Cl
67-66-3, 513-35-9
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
ClC(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY
Melting Point -63.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 280nm, 0.01 max. at 270nm, 0.075 max. at 260nm, 0.5 max. at 250nm, 1 max. at 245nm
Acidity 0.0002 meq/g max.
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.0005% max.
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Beilstein 01, 61
Water 0.05% max.
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
IUPAC Name chloroform
Formula Weight 119.38
Grade HPLC
Chemical Name or Material Chloroform, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with amylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'