missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl phenyl sulfone, 99%, Thermo Scientific Chemicals

Catalog Number p-4089728
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7205-98-3
C7H7ClO2S
190.65
MFCD00007551
NXAIQSVCXQZNRY-UHFFFAOYSA-N
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
81625
chloromethylsulfonylbenzene
C1=CC=C(C=C1)S(=O)(=O)CCl
This item is not returnable. View return policy
7205-98-3
C7H7ClO2S
190.65
MFCD00007551
NXAIQSVCXQZNRY-UHFFFAOYSA-N
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
81625
chloromethylsulfonylbenzene
C1=CC=C(C=C1)S(=O)(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 51°C to 53°C
Flash Point 113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SO2CH2Cl
Quantity 5 g
Formula Weight 190.65
Percent Purity 99%
Chemical Name or Material Chloromethyl phenyl sulfone
compliance-icons
Product Identifier
  • Chloromethyl phenyl sulfone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'