missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

Catalog Number p-4510144
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
13154-24-0
C9H21ClSi
192.80
MFCD00009656
KQIADDMXRMTWHZ-UHFFFAOYSA-N
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
139400
CC(C)[Si](Cl)(C(C)C)C(C)C
13154-24-0
C9H21ClSi
192.80
MFCD00009656
KQIADDMXRMTWHZ-UHFFFAOYSA-N
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
139400
CC(C)[Si](Cl)(C(C)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9020g/mL
Boiling Point 198.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]3SiCl
Refractive Index 1.4520 to 1.4540
Quantity 10 g
Specific Gravity 0.902
Solubility Information Solubility in water: reacts
IUPAC Name chloro-tri(propan-2-yl)silane
Formula Weight 192.81
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chlorotriisopropylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorotriisopropylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'