Learn More
Choice™ Polypropylene (PP) Syringe Filters
Use Choice Polypropylene Syringe Filters as an economic solution to filtration of biological samples, and for filtration of aggressive organic solutions.
71.99 € - 82.11 €
Specifications
Disposable | Single-use |
---|---|
Material | Polypropylene |
Material (Membrane) | Polypropylene |
Sterility | Non-sterile |
For Use With (Application) | HPLC, SPE |
Product Code | Brand | Pore Size | Diameter (Metric) | Prefilter | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Prefilter | Price | Quantity & Availability | |||
17715034
|
Thermo Scientific
CH2225-PP |
0.2 μm | 25 mm | No |
82.11 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
17814861
|
Thermo Scientific
CH4525-PP |
0.45 μm | 25 mm | No |
82.11 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
17894851
|
Thermo Scientific
CH2213-PP |
0.2 μm | 13 mm | No |
73.45 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
17804861
|
Thermo Scientific
CH4513-PP |
0.45 μm | 13 mm | No |
71.99 €
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
Description
Thermo Scientific™ Choice™ Polypropylene Syringe Filters are a good choice for filtration of biological samples, and for filtration of aggressive organic solutions. They provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.
Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect instruments and prolong column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.
Specifications- Membrane: Hydrophilic polypropylene
- Maximum operating temperature: 110 °C
- Housing: Medical grade, virgin polypropylene
- Protein- or peptide-based assays
- General HPLC analysis
- Hexane, toluene, benzene, limited resistance to dichloromethane and chloroform
Specifications
Single-use | |
Polypropylene | |
HPLC, SPE | |
100 | |
90 psi | |
Syringe Filter |
Polypropylene | |
Non-sterile | |
Solvent Resistent, Low-Extractable Polypropylene | |
No | |
Choice |