missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride, 99%, Thermo Scientific Chemicals

Catalog Number 10011973
Click to view available options
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67-48-1
C5H14ClNO
139.62
MFCD00011721
SGMZJAMFUVOLNK-UHFFFAOYSA-M
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
6209
CHEBI:133341
2-hydroxyethyl(trimethyl)azanium;chloride
[Cl-].C[N+](C)(C)CCO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67-48-1
C5H14ClNO
139.62
MFCD00011721
SGMZJAMFUVOLNK-UHFFFAOYSA-M
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
6209
CHEBI:133341
2-hydroxyethyl(trimethyl)azanium;chloride
[Cl-].C[N+](C)(C)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 298.0°C to 304.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Argentometry)
Packaging Plastic bottle
Linear Formula HOCH2CH2N(CH3)3Cl
Quantity 50 g
Beilstein 04, II, 722
Merck Index 15, 2211
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 139.62
Percent Purity 99%
Physical Form Adhering Crystals
Chemical Name or Material Choline chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'