missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ HR-P Glass SPE Column

Brand:  Macherey-Nagel™ 730108G

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10022971

  • 192.00 € / Pack of 30

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

3 mL
300mg
SPE Column
SPE
50 to 100 μm
1200 m2/g
Videos
SDS
Documents
Certificates
Special Offers

Special Offers