missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromabond™ HR-XC Polypropylene SPE Column
Brand: Macherey-Nagel™ 730969
This item is not returnable.
View return policy
Specifications
PS/DVB Copolymer | |
SPE | |
Solid Phase Extraction | |
Chromabond HR-XC | |
Spherical | |
800 m2/g |
1 mL | |
30 mg | |
85 μm | |
Solid Phase Extraction Column | |
65 to 75 Å |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction