Learn More
Macherey Nagel SPE method development kit CHROMABOND polymer 5 columns with 200mg each of HR-X, HR-XC (MCX) HR-XA (MAX), HR-P, Easy, PS-H+, PS-OH- 3mL for investigating clean-up separation mechanisms Macherey-Nagel
Brand:  Macherey Nagel 730288
SPE method development kit CHROMABOND polymer 5 columns with 200mg each of HR-X, HR-XC (MCX) HR-XA (MAX), HR-P, Easy, PS-H+, PS-OH- 3mL for investigating clean-up separation mechanisms Macherey-Nagel
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 12406626