missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket

Brand:  Lenz Laborglasinstrumente™ 05450701

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10030511

  • 58.20 € / Each
Estimated Shipment: 20-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Chromatographic
Chromatographic applications
15 mL
Duran Glass
0 to 2.5mm Bore PTFE-Hahn Stopcock
10 mm
Chromatography Column
200 mm
10 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers