Learn More
Chromium(Vi) Oxide, Technical, Fisher Chemical™
Brand: Fisher Chemical C/5840/53
Code : 21
Additional Details : CAS Number : 1333-82-0 Weight : 0.50000kg Transport : UN number : 1463 Chem class : 5.1 Pack group : II
Specifications
1333-82-0 | |
10952 | |
Solid | |
Red to Violet | |
1 |
CrO3 | |
99.99g/mol | |
Plastic powder jar | |
196°C | |
500g |