missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A / CHGA (Neuroendocrine Marker) Mouse, Clone: rCHGA/798, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1113-MSM17-P1
Additional Details : Weight : 0.01000kg
Specifications
Chromogranin A / CHGA (Neuroendocrine Marker) | |
Finely granular cytoplasmic | |
rCHGA/798 | |
Unconjugated | |
Mouse | |
68-75kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
1113 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human CHGA protein | |
PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |