missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Description
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.
Solubility
Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.
Solubility
Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Type | Ciprofloxacin |
Physical Form | Powder |
Melting Point | 253°C to 257°C |
Beilstein | 3568352 |
Merck Index | 14,2314 |
Solubility Information | Soluble in 0;1N HCl at 25mg/ml; Poorly soluble in DMSO |
Formula Weight | 331.3 |
Percent Purity | 98% |
Color | White |
Quantity | 25 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'