missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanediol, 99%, Thermo Scientific Chemicals

Catalog Number 10003450
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1792-81-0
C6H12O2
116.16
MFCD00064944
PFURGBBHAOXLIO-OLQVQODUSA-N
cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
92903
CHEBI:32329
(1R,2S)-cyclohexane-1,2-diol
C1CCC(C(C1)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1792-81-0
C6H12O2
116.16
MFCD00064944
PFURGBBHAOXLIO-OLQVQODUSA-N
cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
92903
CHEBI:32329
(1R,2S)-cyclohexane-1,2-diol
C1CCC(C(C1)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 101°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 1 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 116.16
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material cis-1, 2-Cyclohexanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'