missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Catalog Number p-4510489
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
136315-70-3
C11H19NO4
229.28
BUEPEVBYNBQNED-SFYZADRCSA-N
1r,2s-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-boc-amino cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-1-cyclopentanecarboxylic acid, cis-2-bocamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, +/--cis-2-boc-amino cyclopentanecarboxylic acid
1268160
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCCC1C(=O)O
This item is not returnable. View return policy
136315-70-3
C11H19NO4
229.28
BUEPEVBYNBQNED-SFYZADRCSA-N
1r,2s-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-boc-amino cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-1-cyclopentanecarboxylic acid, cis-2-bocamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, +/--cis-2-boc-amino cyclopentanecarboxylic acid
1268160
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCCC1C(=O)O
Melting Point 90.0°C to 94.0°C
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 229.28
Percent Purity 98%
Quantity 250 mg
Physical Form Fine Crystalline Powder
Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'