missing translation for 'onlineSavingsMsg'
Learn More

Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-93213
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
5392-40-5
C10H16O
152.24
MFCD00006997
WTEVQBCEXWBHNA-JXMROGBWSA-N
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
638011
CHEBI:16980
(2E)-3,7-dimethylocta-2,6-dienal
CC(C)=CCC\C(C)=C\C=O
This item is not returnable. View return policy
5392-40-5
C10H16O
152.24
MFCD00006997
WTEVQBCEXWBHNA-JXMROGBWSA-N
citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
638011
CHEBI:16980
(2E)-3,7-dimethylocta-2,6-dienal
CC(C)=CCC\C(C)=C\C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -10°C
Color Yellow
Density 0.8890g/mL
Boiling Point 229°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCHO
Refractive Index 1.4860 to 1.49
Quantity 5 mL
Merck Index 15, 2321
Specific Gravity 0.889
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 5 volumes of 60% alcohol,soluble in al,benzyl benzoate,diethyl phthalate,glycerol,prop,mineral oil,fixed oils
Formula Weight 152.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Citral
Show More Show Less
compliance-icons
Product Identifier
  • Citral, mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'