missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, for Amino Acid Analysis, Fisher Chemical™

Catalog Number 10020050
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
5949-29-1
210.14
149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
This item is not returnable. View return policy
5949-29-1
210.14
149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
pH 1.8
Melting Point 135°C
Color White
Molecular Formula C6H10O8
Formula Weight 210.15g/mol
Quantity 1 kg
Packaging Plastic powder jar
Physical Form Solid
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'