missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, for Amino Acid Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Specifications
pH | 1.8 |
Melting Point | 135°C |
Color | White |
Molecular Formula | C6H8O7·H2O |
Formula Weight | 210.15g/mol |
Quantity | 500 g |
Packaging | Plastic powder jar |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'