missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation

Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR™ glass.

Brand:  Hirschmann™ 1340140

Additional Details : Weight : 0.19440kg

 View more versions of this product

Product Code. 11787991

  • 82.20 € / Pack of 12
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • One mark
  • Acc. to DIN EN ISO 648
  • Conformity certified, with dated batch identification
Specifications

Specifications

AS
Yes
12/Pk.
2 mL
Volumetric Pipet
AR-GLAS
Brown
0.01 mL
350 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers